siRNA Details
| Gene Symbol: NRIP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020645.2 | NP_065696.1 | 7 | 1270 | ||
| GenBank mRNA | AK024750.1 | - | 7 | 1223 | ||
| AK098694.1 | - | 7 | 1223 | |||
| BC008835.2 | - | 7 | 1204 | 4304488 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

NRIP3| Gene Symbol: NRIP3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_020645.2 | NP_065696.1 | 7 | 1270 | ||
| GenBank mRNA | AK024750.1 | - | 7 | 1223 | ||
| AK098694.1 | - | 7 | 1223 | |||
| BC008835.2 | - | 7 | 1204 | 4304488 | ||