siRNA Details
| Gene Symbol: CD209 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001144893.1 | NP_001138365.1 | 5 | 2815 | ||
| NM_001144894.1 | NP_001138366.1 | 5 | 3091 | |||
| NM_001144895.1 | NP_001138367.1 | 7 | 2947 | |||
| NM_001144896.1 | NP_001138368.1 | 6 | 3151 | |||
| NM_001144897.1 | NP_001138369.1 | 7 | 3205 | |||
| NM_001144899.1 | NP_001138371.1 | 7 | 2740 | |||
| NM_021155.3 | NP_066978.1 | 7 | 3223 | |||
| NR_026692.1 | 6 | 3323 | ||||
| XM_005272472.4 | XP_005272529.1 | 7 | 2840 | |||
| GenBank mRNA | AF290886.1 | - | 7 | 3164 | ||
| AK293089.1 | - | 7 | 3178 | |||

CD209
Genomic Map