siRNA Details
Gene Symbol: ZNF462 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_021224.4 | NP_067047.4 | 3 | 4831 | ||
XM_006717209.3 | XP_006717272.1 | 3 | 4827 | |||
XM_006717211.3 | XP_006717274.1 | 3 | 5873 | |||
XM_006717212.3 | XP_006717275.1 | 3 | 4632 | |||
XM_006717215.3 | XP_006717278.1 | 3 | 6622 | |||
XM_006717216.3 | XP_006717279.1 | 3 | 6617 | |||
XM_011518892.2 | XP_011517194.1 | 3 | 6622 | |||
XM_017014996.1 | XP_016870485.1 | 3 | 4974 | |||
XM_017014997.1 | XP_016870486.1 | 3 | 4789 | |||
GenBank mRNA | AB058706.2 | - | 1 | 1077 | ||
AK027866.1 | - | 1 | 1191 | |||
BX648965.1 | - | 3 | 4831 | |||
CR749850.1 | - | 1 | 4233 |

More Information
Other Species Targeted

This siRNA also targets the following transcripts in Mouse: NM_172867, DQ355518.
Panther Classification:
Gene Ontology Categories:
