siRNA Details
| Gene Symbol: DMRT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021240.3 | NP_067063.1 | 2 | 1766 | ||
| GenBank mRNA | AF193873.1 | - | 2 | 1347 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DMRT3| Gene Symbol: DMRT3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021240.3 | NP_067063.1 | 2 | 1766 | ||
| GenBank mRNA | AF193873.1 | - | 2 | 1347 | ||