siRNA Details
| Gene Symbol: TRIM39 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021253.3 | NP_067076.2 | 9 | 2866 | ||
| NM_172016.2 | NP_742013.1 | 8 | 3090 | |||
| GenBank mRNA | AX776007.1 | - | 8 | 2656 | ||
| BC007661.2 | - | 8 | 2647 | 3051389 | ||
| BC034985.1 | - | 8 | 3063 | 5268723 | ||

TRIM39
Genomic Map