siRNA Details
| Gene Symbol: GOLGA1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002077.3 | NP_002068.1 | 3, 4 | 474 | ||
| XM_005251929.3 | XP_005251986.1 | 3, 4 | 406 | |||
| XM_006717062.3 | XP_006717125.1 | 3, 4 | 470 | |||
| XM_006717063.3 | XP_006717126.1 | 3, 4 | 441 | |||
| GenBank mRNA | AK307789.1 | - | 1, 2 | 216 | ||
| BC032853.1 | - | 3, 4 | 444 | 4828793 | ||
| BG699932.1 | - | 3, 4 | 447 | |||
| U51587.1 | - | 3, 4 | 466 | |||

GOLGA1
Genomic Map