siRNA Details
| Gene Symbol: FGFRL1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001004356.2 | NP_001004356.1 | 7 | 2694 | ||
| NM_001004358.1 | NP_001004358.1 | 7 | 2577 | |||
| NM_021923.3 | NP_068742.2 | 6 | 2567 | |||
| XM_011513486.1 | XP_011511788.1 | 7 | 2698 | |||
| GenBank mRNA | AF312678.1 | - | 2878 | |||
| AJ277437.1 | - | 6 | 2567 | |||
| AK000530.1 | - | 1 | 232 | |||
| AK022597.1 | - | 1 | 1479 | |||
| AY358303.1 | - | 2899 | ||||
| BC015228.1 | - | 3 | 1978 | |||
| BC036769.1 | - | 7 | 2706 | 5744684 | ||

FGFRL1
Genomic Map