siRNA Details
| Gene Symbol: ZNF148 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_021964.2 | NP_068799.2 | 9 | 2565 | ||
| GenBank mRNA | AF039019.1 | - | 8 | 2290 | ||
| AF432210.1 | - | 9 | 2381 | |||
| AJ236885.1 | - | 9 | 2470 | |||
| AK314203.1 | - | 9 | 2500 | |||
| AL833616.1 | - | 9 | 2403 | |||
| BC050260.1 | - | 10 | 2641 | 4423572 | ||

ZNF148
Genomic Map