siRNA Details
| Gene Symbol: SLC13A3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001011554.2 | NP_001011554.1 | 6 | 750 | ||
| NM_001193339.1 | NP_001180268.1 | 4 | 647 | |||
| NM_001193340.1 | NP_001180269.1 | 6 | 750 | |||
| NM_001193342.1 | NP_001180271.1 | 5 | 541 | |||
| NM_022829.5 | NP_073740.2 | 5 | 714 | |||
| GenBank mRNA | AF154121.1 | - | 5 | 693 | ||
| AK056713.1 | - | 5 | 712 | |||
| AK096658.1 | - | 6 | 750 | |||
| AK293902.1 | - | 5 | 530 | |||
| AK295748.1 | - | 4 | 645 | |||
| AK310960.1 | - | 4 | 724 | |||
| AK315131.1 | - | 5 | 712 | |||
| AY072810.3 | - | 5 | 693 | |||
| BC035966.1 | - | 5 | 712 | 4609776 | ||
| BM474324.1 | - | 145 | ||||
| CB997336.1 | - | 5 | 566 | |||

SLC13A3
Genomic Map