siRNA Details
| Gene Symbol: ZNF426 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001300883.2 | NP_001287812.1 | 6 | 1289 | ||
| NM_001318055.1 | NP_001304984.1 | 7 | 1025 | |||
| NM_001318056.1 | NP_001304985.1 | 7 | 1069 | |||
| NM_024106.2 | NP_077011.1 | 8 | 1152 | |||
| XM_017027293.1 | XP_016882782.1 | 7 | 1088 | |||
| XM_017027294.1 | XP_016882783.1 | 5 | 1194 | |||
| GenBank mRNA | AB209837.1 | - | 6 | 1234 | ||
| AK095759.1 | - | 6 | 1207 | |||
| BC001791.1 | - | 8 | 1134 | 3543910 | ||

ZNF426
Genomic Map