siRNA Details
| Gene Symbol: ZNF665 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024733.3 | NP_079009.3 | 4 | 3053 | ||
| GenBank mRNA | AL832507.1 | - | 4 | 3037 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

ZNF665| Gene Symbol: ZNF665 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024733.3 | NP_079009.3 | 4 | 3053 | ||
| GenBank mRNA | AL832507.1 | - | 4 | 3037 | ||