siRNA Details
| Gene Symbol: GRHL2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024915.3 | NP_079191.2 | 16 | 2195 | ||
| XM_011517305.2 | XP_011515607.1 | 16 | 2226 | |||
| XM_011517306.2 | XP_011515608.1 | 16 | 1963 | |||
| GenBank mRNA | AK002034.1 | - | 3 | 219 | ||
| AK023844.1 | - | 16 | 1877 | |||
| AK292853.1 | - | 16 | 1877 | |||
| BC069618.1 | - | 16 | 1877 | 7262389 | ||
| BC069633.1 | - | 16 | 1937 | 7262401 | ||
| BC069638.1 | - | 16 | 1877 | 7262413 | ||
| BC129822.1 | - | 16 | 1877 | 40113571 | ||
| BC129823.1 | - | 16 | 1877 | 40113576 | ||

GRHL2
Genomic Map