siRNA Details
| Gene Symbol: NUP210 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024923.3 | NP_079199.2 | 12 | 1646 | ||
| XM_005264994.3 | XP_005265051.1 | 12 | 1646 | |||
| XM_011533538.2 | XP_011531840.1 | 12 | 1646 | |||
| XM_017006021.1 | XP_016861510.1 | 12 | 1646 | |||
| GenBank mRNA | AK026042.1 | - | 3 | 321 | ||
| AK225409.1 | - | 3 | 321 | |||
| BC067089.1 | - | 12 | 1564 | 6281693 | ||

NUP210
Genomic Map