siRNA Details
| Gene Symbol: NUP210 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_024923.3 | NP_079199.2 | 30 | 4085 | ||
| XM_017006018.1 | XP_016861507.1 | 12 | 1918 | |||
| XM_017006019.1 | XP_016861508.1 | 12 | 1885 | |||
| XM_017006020.1 | XP_016861509.1 | 12 | 3957 | |||
| GenBank mRNA | AB020713.1 | - | 10 | 1101 | ||
| AK074101.1 | - | 15 | 1707 | |||

NUP210
Genomic Map