siRNA Details
| Gene Symbol: FOXRED2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001102371.1 | NP_001095841.1 | 4 | 982 | ||
| NM_024955.5 | NP_079231.4 | 4 | 1021 | |||
| GenBank mRNA | AK025714.1 | - | 4 | 921 | ||
| AK225347.1 | - | 4 | 921 | |||
| AK315555.1 | - | 4 | 924 | |||
| BC015726.1 | - | 4 | 905 | 4866761 | ||
| BC027716.2 | - | 4 | 1000 | 4127473 | ||
| BC040351.1 | - | 4 | 944 | 3873448 | ||
| CR456444.1 | - | 3 | 996 | |||

FOXRED2
Genomic Map