siRNA Details
Gene Symbol: FOXRED2 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001102371.1 | NP_001095841.1 | 4 | 1062 | ||
NM_024955.5 | NP_079231.4 | 4 | 1101 | |||
GenBank mRNA | AK025714.1 | - | 4 | 1001 | ||
AK225347.1 | - | 4 | 1001 | |||
AK315555.1 | - | 4 | 1004 | |||
BC015726.1 | - | 4 | 985 | 4866761 | ||
BC027716.2 | - | 4 | 1080 | 4127473 | ||
BC040351.1 | - | 4 | 1024 | 3873448 | ||
CR456444.1 | - | 3 | 1076 |

More Information
Other Species Targeted

Panther Classification:
Gene Ontology Categories:
