siRNA Details
| Gene Symbol: FOXRED2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001102371.1 | NP_001095841.1 | 9 | 2756 | ||
| NM_024955.5 | NP_079231.4 | 9 | 2795 | |||
| XM_017028941.1 | XP_016884430.1 | 8 | 2428 | |||
| XM_017028942.1 | XP_016884431.1 | 8 | 2525 | |||
| GenBank mRNA | AK025714.1 | - | 9 | 2695 | ||
| AK026975.1 | - | 6 | 1876 | |||
| AK091053.1 | - | 4 | 1687 | |||
| BC015726.1 | - | 9 | 2679 | 4866761 | ||
| BC027716.2 | - | 9 | 2774 | 4127473 | ||
| BC040351.1 | - | 9 | 2718 | 3873448 | ||

FOXRED2
Genomic Map