siRNA Details
| Gene Symbol: PROSER1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025138.4 | NP_079414.3 | 13 | 4598 | ||
| XM_005266545.3 | XP_005266602.1 | 12 | 4571 | |||
| XM_011535239.2 | XP_011533541.1 | 12 | 4531 | |||
| GenBank mRNA | AB107354.1 | - | 13 | 4598 | ||
| AK024308.1 | - | 3 | 1867 | |||
| AL833854.1 | - | 3 | 2325 | |||
| BC064348.1 | - | 13 | 3853 | 4939198 | ||
| BX647471.1 | - | 13 | 4020 | |||

PROSER1
Genomic Map