siRNA Details
| Gene Symbol: PDGFD | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_025208.4 | NP_079484.1 | 6 | 1204 | ||
| NM_033135.3 | NP_149126.1 | 6 | 1186 | |||
| GenBank mRNA | AB033832.1 | - | 6 | 988 | ||
| AF113216.1 | - | 6 | 938 | |||
| AF335584.1 | - | 6 | 1013 | |||
| AF336376.1 | - | 6 | 1007 | |||
| AK292801.1 | - | 6 | 1012 | |||
| AY027517.1 | - | 6 | 924 | |||
| AY027518.1 | - | 6 | 906 | |||
| AY359116.1 | - | 6 | 845 | |||
| BC030645.2 | - | 6 | 1266 | 4824526 | ||

PDGFD
Genomic Map