siRNA Details
| Gene Symbol: PRR7 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001174101.1 | NP_001167572.1 | 3 | 1380 | ||
| NM_001174102.1 | NP_001167573.1 | 3 | 1297 | |||
| NM_030567.4 | NP_085044.2 | 4 | 1465 | |||
| XM_011534662.1 | XP_011532964.1 | 3 | 7203 | |||
| XM_011534663.2 | XP_011532965.1 | 3 | 1941 | |||
| XM_017009896.1 | XP_016865385.1 | 2 | 7991 | |||
| XM_017009897.1 | XP_016865386.1 | 3 | 2173 | |||
| GenBank mRNA | BC004261.1 | - | 4 | 1297 | 3606982 | |
| BC021240.1 | - | 3 | 1288 | 3603954 | ||
| BC024233.2 | - | 4 | 1469 | 4697841 | ||
| BC033649.1 | - | 2 | 1053 | 5538503 | ||

PRR7
Genomic Map