siRNA Details
| Gene Symbol: TMX1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_030755.4 | NP_110382.3 | 8 | 1881 | ||
| GenBank mRNA | AK075395.1 | - | 8 | 1783 | ||
| AL080080.1 | - | 8 | 1779 | |||
| AY358640.1 | - | 8 | 1763 | |||
| BC036460.2 | - | 8 | 1830 | 4814497 | ||
| BC056874.1 | - | 8 | 1770 | 5740203 | ||

TMX1
Genomic Map