siRNA Details
| Gene Symbol: TEX101 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001130011.1 | NP_001123483.1 | 6 | 1051 | ||
| NM_031451.4 | NP_113639.4 | 9 | 1471 | |||
| XM_005259303.3 | XP_005259360.2 | 6 | 1041 | |||
| GenBank mRNA | AF241268.1 | - | 9 | 1470 | ||
| AF353396.1 | - | 6 | 1141 | |||
| AY014285.1 | - | 6 | 1023 | |||
| AY359077.1 | - | 6 | 1019 | |||
| BC001861.2 | - | 7 | 1218 | 3538494 | ||

TEX101
Genomic Map