siRNA Details
| Gene Symbol: GRWD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_031485.3 | NP_113673.3 | 7 | 1661 | ||
| GenBank mRNA | AF337808.1 | - | 7 | 1440 | ||
| AK074676.1 | - | 7 | 1629 | |||
| AK131055.1 | - | 6 | 867 | |||
| AK300225.1 | - | 7 | 1114 | |||
| AL713729.1 | - | 7 | 1436 | |||
| BC002440.2 | - | 7 | 1436 | 3347366 | ||
| BC036281.1 | - | 7 | 1412 | |||
| EF011614.1 | - | 7 | 1434 | |||

GRWD1
Genomic Map