siRNA Details
| Gene Symbol: ZIC4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001168378.1 | NP_001161850.1 | 5 | 1718 | ||
| NM_001168379.1 | NP_001161851.1 | 5 | 1545 | |||
| NM_001243256.1 | NP_001230185.1 | 4 | 1226 | |||
| NM_032153.5 | NP_115529.2 | 5 | 1844 | |||
| NR_033118.2 | 3 | 1155 | ||||
| NR_033119.2 | 3 | 1101 | ||||
| NR_040762.1 | 3 | 1002 | ||||
| GenBank mRNA | AK056262.1 | - | 5 | 1655 | ||
| AK056394.1 | - | 3 | 2029 | |||
| AK094286.1 | - | 3 | 1341 | |||
| AK124600.1 | - | 1 | 2180 | |||
| AK126595.1 | - | 2 | 2381 | |||
| AK127733.1 | - | 3 | 1117 | |||
| AK294831.1 | - | 5 | 1718 | |||
| BC126267.1 | - | 5 | 1480 | 8991983 | ||
| BC136339.1 | - | 5 | 1480 | 9020326 | ||

ZIC4
Genomic Map