siRNA Details
| Gene Symbol: RHOXF2B | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001099685.1 | NP_001093155.1 | 4 | 779 | ||
| GenBank mRNA | AL590524.1 | - | 4 | 675 | ||
| Gene Symbol: RHOXF2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_032498.2 | NP_115887.1 | 4 | 779 | ||
| GenBank mRNA | AF317219.1 | - | 4 | 719 | ||
| AK058125.1 | - | 4 | 672 | |||
| AL590526.1 | - | 4 | 779 | |||
| BC021719.2 | - | 4 | 674 | 5269333 | ||

RHOXF2B
Genomic Map