siRNA Details
| Gene Symbol: RTN4IP1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001318746.1 | NP_001305675.1 | 6 | 633 | ||
| NM_032730.5 | NP_116119.2 | 6 | 1219 | |||
| XM_011536192.2 | XP_011534494.1 | 7 | 1307 | |||
| XM_017011375.1 | XP_016866864.1 | 6 | 1219 | |||
| XM_017011376.1 | XP_016866865.1 | 6 | 1219 | |||
| GenBank mRNA | AK095207.1 | - | 6 | 633 | ||
| AK313962.1 | - | 6 | 1137 | |||
| AY063761.1 | - | 6 | 742 | |||
| BC006399.2 | - | 6 | 1014 | 4309263 | ||

RTN4IP1
Genomic Map