siRNA Details
| Gene Symbol: AIFM2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001198696.1 | NP_001185625.1 | 9 | 2446 | ||
| NM_032797.5 | NP_116186.1 | 9 | 2443 | |||
| GenBank mRNA | AK027403.1 | - | 9 | 2322 | ||
| AK127353.1 | - | 9 | 4198 | |||
| BX537621.1 | - | 6 | 3080 | |||

AIFM2
Genomic Map