siRNA Details
| Gene Symbol: ZNF382 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001256838.1 | NP_001243767.1 | 5 | 2130 | ||
| NM_032825.4 | NP_116214.2 | 5 | 2155 | |||
| GenBank mRNA | AF513816.1 | - | 5 | 2135 | ||
| AK027592.1 | - | 5 | 2135 | |||
| AL832265.1 | - | 3 | 3519 | |||
| AY227449.1 | - | 5 | 2135 | |||
| BC038358.1 | - | 4 | 2592 | |||
| BC132675.1 | - | 4 | 2100 | 40146697 | ||

ZNF382
Genomic Map