siRNA Details
Gene Symbol: RELT | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_032871.3 | NP_116260.2 | 11 | 2665 | ||
NM_152222.1 | NP_689408.1 | 11 | 2603 | |||
XM_005274356.2 | XP_005274413.1 | 6 | 2766 | |||
XM_006718714.2 | XP_006718777.1 | 9 | 2230 | |||
XM_011545305.1 | XP_011543607.1 | 10 | 2488 | |||
GenBank mRNA | BC051810.1 | - | 11 | 2554 | 5216988 |
