siRNA Details
| Gene Symbol: TRIM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF220023.1 | - | 7 | 978 | ||
| AF220024.1 | - | 5 | 900 | |||
| AK092892.1 | - | 5 | 925 | |||
| AK293565.1 | - | 7 | 817 | |||
Please enter the information below and press OK to send your cart to Core Services for purchase.

TRIM4| Gene Symbol: TRIM4 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| GenBank mRNA | AF220023.1 | - | 7 | 978 | ||
| AF220024.1 | - | 5 | 900 | |||
| AK092892.1 | - | 5 | 925 | |||
| AK293565.1 | - | 7 | 817 | |||