siRNA Details
| Gene Symbol: ZNF101 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001300949.1 | NP_001287878.1 | 4 | 852 | ||
| NM_033204.3 | NP_149981.2 | 4 | 980 | |||
| GenBank mRNA | AK097169.1 | - | 4 | 972 | ||
| AL096747.1 | - | 3 | 2525 | |||
| AY149174.1 | - | 4 | 914 | |||
| BC011502.1 | - | 1 | 616 | |||
| BC094808.1 | - | 4 | 913 | |||
| BC119680.1 | - | 5 | 1016 | 40116294 | ||

ZNF101
Genomic Map