siRNA Details
| Gene Symbol: CSMD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033225.5 | NP_150094.5 | 9 | 1544 | ||
| XM_011534752.2 | XP_011533054.1 | 9 | 1773 | |||
| XM_017013731.1 | XP_016869220.1 | 9 | 1773 | |||
| GenBank mRNA | AF333704.2 | - | 10 | 1547 | ||
| AY017307.1 | - | 10 | 1442 | |||
| CD634856.1 | - | 2 | 89 | |||

CSMD1
Genomic Map