siRNA Details
| Gene Symbol: CSMD1 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033225.5 | NP_150094.5 | 23 | 4008 | ||
| XM_011534752.2 | XP_011533054.1 | 23 | 4237 | |||
| XM_011534753.2 | XP_011533055.1 | 6 | 1115 | |||
| XM_017013731.1 | XP_016869220.1 | 23 | 4237 | |||
| GenBank mRNA | AB067477.1 | - | 4 | 726 | ||
| AB209502.1 | - | 11 | 2060 | |||
| AF333704.2 | - | 24 | 4011 | |||
| AK126857.1 | - | 14 | 2180 | |||
| AY017307.1 | - | 24 | 3906 | |||
| AY358174.1 | - | 7 | 1151 | |||

CSMD1
Genomic Map