siRNA Details
| Gene Symbol: BTBD6 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_033271.2 | NP_150374.2 | 5 | 783 | ||
| XM_005268188.3 | XP_005268245.1 | 4 | 951 | |||
| GenBank mRNA | AF173358.1 | - | 1 | 528 | ||
| AF353674.1 | - | 4 | 647 | |||
| BC042525.1 | - | 5 | 753 | 4824525 | ||

BTBD6
Genomic Map