siRNA Details
| Gene Symbol: PGLYRP2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_052890.3 | NP_443122.3 | 5 | 1805 | ||
| GenBank mRNA | AB073610.1 | - | 5 | 1754 | ||
| AF384856.1 | - | 5 | 1698 | |||
| AK055882.1 | - | 4 | 2681 | |||
| AK289415.1 | - | 5 | 1738 | |||
| AK292292.1 | - | 5 | 2153 | |||
| AY358156.1 | - | 5 | 1729 | |||

PGLYRP2
Genomic Map