siRNA Details
| Gene Symbol: FCRL3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001320333.1 | NP_001307262.1 | 5 | 720 | ||
| NM_052939.3 | NP_443171.2 | 5 | 705 | |||
| NR_135214.1 | 5 | 720 | ||||
| NR_135215.1 | 5 | 720 | ||||
| NR_135216.1 | 5 | 720 | ||||
| NR_135217.1 | 5 | 720 | ||||
| XM_006711145.1 | XP_006711208.1 | 5 | 635 | |||
| GenBank mRNA | AF416901.1 | - | 5 | 584 | ||
| AF416902.1 | - | 5 | 584 | |||
| AF416903.1 | - | 5 | 584 | |||
| AF416904.1 | - | 5 | 584 | |||
| AF416905.1 | - | 5 | 584 | |||
| AF459027.1 | - | 5 | 705 | |||
| AK098122.1 | - | 5 | 635 | |||
| AK301257.1 | - | 5 | 607 | |||
| AL833391.1 | - | 1 | 209 | |||
| AY043466.1 | - | 4 | 509 | |||
| BC028933.1 | - | 5 | 586 | 4340939 | ||

FCRL3
Genomic Map