siRNA Details
| Gene Symbol: SLC26A8 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001193476.1 | NP_001180405.1 | 3, 4 | 447 | ||
| NM_052961.3 | NP_443193.1 | 3, 4 | 471 | |||
| NM_138718.2 | NP_619732.2 | 3, 4 | 509 | |||
| XM_011514294.2 | XP_011512596.1 | 3, 4 | 690 | |||
| XM_017010235.1 | XP_016865724.1 | 3, 4 | 497 | |||
| GenBank mRNA | AF314959.1 | - | 2, 3 | 341 | ||
| AF331522.1 | - | 3, 4 | 415 | |||
| AK122928.1 | - | 3, 4 | 447 | |||
| AK125189.1 | - | 3, 4 | 691 | |||
| BC025408.2 | - | 3, 4 | 502 | 4823098 | ||

SLC26A8
Genomic Map