siRNA Details
| Gene Symbol: ABCA10 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080282.3 | NP_525021.3 | 16 | 2600 | ||
| GenBank mRNA | AL832004.2 | - | 16 | 2306 | ||
| AY028900.1 | - | 16 | 2600 | |||
| AY247065.1 | - | 16 | 2631 | |||
| BC136460.1 | - | 13 | 1805 | |||
| BC144124.1 | - | 13 | 1874 | |||

ABCA10
Genomic Map