siRNA Details
| Gene Symbol: DCBLD2 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_080927.3 | NP_563615.3 | 10 | 1613 | ||
| XM_011512419.2 | XP_011510721.1 | 9 | 1329 | |||
| GenBank mRNA | AB073146.1 | - | 10 | 1613 | ||
| AF387547.1 | - | 10 | 1261 | |||
| AK056350.1 | - | 10 | 1606 | |||
| AK302649.1 | - | 10 | 1093 | |||
| BC029658.1 | - | 10 | 1132 | 4431275 | ||
| BX647140.1 | - | 1 | 3846 | |||

DCBLD2
Genomic Map