siRNA Details
| Gene Symbol: GRIN3A | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_133445.2 | NP_597702.2 | 9 | 4659 | ||
| GenBank mRNA | AB075853.1 | - | 9 | 4659 | ||
| BC132866.1 | - | 9 | 4485 | 40146888 | ||
| BC132868.1 | - | 9 | 4485 | 40146890 | ||
| BC167432.1 | - | 9 | 4659 | 8860109 | ||

GRIN3A
Genomic Map