siRNA Details
Gene Symbol: ZFP82 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001321917.1 | NP_001308846.1 | 5 | 2238 | ||
NM_133466.3 | NP_597723.1 | 5 | 2235 | |||
GenBank mRNA | AB075828.1 | - | 1 | 3702 | ||
AK074942.1 | - | 5 | 2217 | |||
AL834267.1 | - | 5 | 2209 | |||
GU388429.1 | - | 5 | 2205 |
