siRNA Details
Gene Symbol: ZNF431 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_001319124.1 | NP_001306053.1 | 5 | 3139 | ||
NM_001319126.1 | NP_001306055.1 | 6 | 3158 | |||
NM_001319127.1 | NP_001306056.1 | 6 | 3216 | |||
NM_133473.2 | NP_597730.2 | 5 | 3109 | |||
XM_011527750.2 | XP_011526052.1 | 6 | 3187 | |||
GenBank mRNA | AB075849.1 | - | 5 | 3071 | ||
BC040506.1 | - | 5 | 3063 | 5297447 |
Gene Symbol: LOC105372321 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
GenBank mRNA | CR627135.1 | - | 1 | 2713 |
Gene Symbol: ZNF714 | ||||||
---|---|---|---|---|---|---|
Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
RefSeq | NM_182515.3 | NP_872321.2 | 5 | 3565 |
