siRNA Details
| Gene Symbol: ZNF274 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001278734.1 | NP_001265663.1 | 8 | 2578 | ||
| NM_016324.3 | NP_057408.2 | 5 | 2262 | |||
| NM_016325.3 | NP_057409.1 | 7 | 2467 | |||
| NM_133502.2 | NP_598009.1 | 8 | 2563 | |||
| XM_011526327.1 | XP_011524629.1 | 7 | 2001 | |||
| XM_011526328.2 | XP_011524630.1 | 3 | 1682 | |||
| XM_017026174.1 | XP_016881663.1 | 6 | 2268 | |||
| XM_017026175.1 | XP_016881664.1 | 6 | 2423 | |||
| GenBank mRNA | AB029149.1 | - | 5 | 2222 | ||
| AB029150.1 | - | 7 | 2369 | |||
| AF275680.1 | - | 8 | 2242 | |||
| AF275681.1 | - | 2 | 1676 | |||
| AK095162.1 | - | 2 | 2907 | |||
| AK222878.1 | - | 5 | 2190 | |||
| BC009763.2 | - | 7 | 2090 | 2966764 | ||
| BX641169.1 | - | 2 | 4677 | |||
| CA942346.1 | - | 1 | 197 | |||
| U71598.1 | - | 1 | 559 | |||

ZNF274
Genomic Map