siRNA Details
| Gene Symbol: GALM | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_138801.2 | NP_620156.1 | 5 | 915 | ||
| GenBank mRNA | AK291489.1 | - | 5 | 723 | ||
| BC014916.1 | - | 5 | 697 | 3920640 | ||
| BC019263.1 | - | 5 | 728 | 3502667 | ||
| EU794611.1 | - | 5 | 697 | |||

GALM
Genomic Map