siRNA Details
| Gene Symbol: RAET1E | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001243325.1 | NP_001230254.1 | 3 | 559 | ||
| NM_001243327.1 | NP_001230256.1 | 3 | 667 | |||
| NM_001243328.1 | NP_001230257.1 | 4 | 881 | |||
| NM_139165.2 | NP_631904.1 | 3 | 667 | |||
| XM_011535475.2 | XP_011533777.1 | 5 | 1187 | |||
| XM_011535476.2 | XP_011533778.1 | 5 | 1201 | |||
| XM_011535478.2 | XP_011533780.1 | 5 | 1184 | |||
| XM_011535479.2 | XP_011533781.1 | 5 | 1186 | |||
| XM_017010285.1 | XP_016865774.1 | 5 | 1186 | |||
| XM_017010286.1 | XP_016865775.1 | 5 | 1186 | |||
| XM_017010287.1 | XP_016865776.1 | 5 | 1186 | |||
| XM_017010288.1 | XP_016865777.1 | 3 | 435 | |||
| GenBank mRNA | AF359243.1 | - | 3 | 555 | ||
| AY054974.1 | - | 3 | 426 | |||
| AY069961.1 | - | 3 | 594 | |||
| AY176317.1 | - | 3 | 534 | |||
| AY252119.1 | - | 3 | 534 | |||
| AY359075.1 | - | 4 | 881 | |||
| BC101976.1 | - | 3 | 459 | |||
| BC103694.1 | - | 3 | 565 | 40003751 | ||
| EF489426.1 | - | 3 | 534 | |||
| KJ628092.1 | - | 2 | 446 | |||

RAET1E
Genomic Map