siRNA Details
| Gene Symbol: ZNF480 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001297624.1 | NP_001284553.1 | 4 | 4136 | ||
| NM_001297625.1 | NP_001284554.1 | 4 | 4138 | |||
| NM_144684.3 | NP_653285.2 | 5 | 4265 | |||
| XM_011526465.2 | XP_011524767.1 | 5 | 4667 | |||
| GenBank mRNA | AL832560.1 | - | 4 | 4115 | ||
| AY512662.1 | - | 5 | 4234 | |||
| BC018783.1 | - | 5 | 4234 | 4873910 | ||

ZNF480
Genomic Map