siRNA Details
| Gene Symbol: AIFM3 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001018060.2 | NP_001018070.1 | 15 | 1565 | ||
| NM_001146288.1 | NP_001139760.1 | 15 | 1650 | |||
| NM_144704.2 | NP_653305.1 | 15 | 1565 | |||
| NR_027464.1 | 14 | 1522 | ||||
| GenBank mRNA | AK055035.1 | - | 15 | 1549 | ||
| AK094844.1 | - | 15 | 1554 | |||
| AK126917.1 | - | 1 | 892 | |||
| AK295581.1 | - | 14 | 1425 | |||
| AK316042.1 | - | 14 | 1418 | |||
| BC032485.1 | - | 15 | 1532 | 5221203 | ||
| CR456342.1 | - | 14 | 1384 | |||

AIFM3
Genomic Map