siRNA Details
| Gene Symbol: DHRSX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145177.2 | NP_660160.2 | 7 | 2520 | ||
| GenBank mRNA | AJ293620.1 | - | 7 | 2479 | ||
Please enter the information below and press OK to send your cart to Core Services for purchase.

DHRSX| Gene Symbol: DHRSX | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_145177.2 | NP_660160.2 | 7 | 2520 | ||
| GenBank mRNA | AJ293620.1 | - | 7 | 2479 | ||