siRNA Details
| Gene Symbol: ZNF396 | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001322286.1 | NP_001309215.1 | 4 | 2018 | ||
| NM_001322290.1 | NP_001309219.1 | 5 | 2255 | |||
| NM_145756.2 | NP_665699.1 | 5 | 1148 | |||
| XM_006722432.3 | XP_006722495.1 | 4 | 2746 | |||
| XM_017025687.1 | XP_016881176.1 | 5 | 2798 | |||
| XM_017025688.1 | XP_016881177.1 | 4 | 2633 | |||
| XM_017025689.1 | XP_016881178.1 | 5 | 947 | |||
| GenBank mRNA | AK055775.1 | - | 5 | 1148 | ||
| BC130288.1 | - | 5 | 1135 | 40146319 | ||
| BC136314.1 | - | 5 | 1135 | 9020301 | ||

ZNF396
Genomic Map