siRNA Details
| Gene Symbol: FNTB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_002028.3 | NP_002019.1 | 12 | 2485 | ||
| GenBank mRNA | AK093298.1 | - | 10 | 2243 | ||
| BC020232.1 | - | 12 | 2335 | 4652202 | ||
| BX248269.1 | - | 12 | 2331 | |||
| Gene Symbol: CHURC1-FNTB | ||||||
|---|---|---|---|---|---|---|
| Interrogated Sequence | Translated Protein | Targeted Exon(s) | siRNA Location | IMAGE Clone ID | ||
| RefSeq | NM_001202558.1 | NP_001189487.1 | 13 | 2486 | ||
| NM_001202559.1 | NP_001189488.1 | 14 | 2557 | |||

FNTB
Genomic Map